Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020947.3 | NP_065998.3 | 3, 4 | 485 | ||
XM_005256074.3 | XP_005256131.1 | 3, 4 | 555 | |||
XM_005256075.2 | XP_005256132.1 | 4, 5 | 683 | |||
XM_006721240.2 | XP_006721303.1 | 4, 5 | 638 | |||
XM_011523248.2 | XP_011521550.1 | 5, 6 | 782 | |||
XM_011523249.2 | XP_011521551.1 | 4, 5 | 633 | |||
XM_017023510.1 | XP_016878999.1 | 5, 6 | 802 | |||
XM_017023511.1 | XP_016879000.1 | 3, 4 | 557 | |||
XM_017023512.1 | XP_016879001.1 | 4, 5 | 725 | |||
GenBank mRNA | AB046829.2 | - | 430 | |||
AK291237.1 | - | 3, 4 | 485 | |||
AK300132.1 | - | 4, 5 | 722 | |||
AK310887.1 | - | 3, 4 | 561 | |||
BC023251.1 | - | 2, 3 | 417 | 4470095 | ||
BC060844.1 | - | 3, 4 | 472 | 30342219 | ||
BI767086.1 | - | 3, 4 | 448 |