Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136557.1 | NP_001130029.1 | 9 | 1064 | ||
NM_001136558.1 | NP_001130030.1 | 9 | 1064 | |||
NM_001287346.1 | NP_001274275.1 | 9 | 1060 | |||
NM_020960.4 | NP_066011.2 | 9 | 1064 | |||
XM_017014953.1 | XP_016870442.1 | 9 | 1025 | |||
XM_017014954.1 | XP_016870443.1 | 9 | 1025 | |||
GenBank mRNA | AB046844.1 | - | 9 | 835 | ||
AF376725.1 | - | 9 | 845 | |||
AK026244.1 | - | 9 | 849 | |||
AK225456.1 | - | 9 | 849 | |||
AK293092.1 | - | 9 | 845 | |||
AL834359.1 | - | 8 | 708 | |||
BC110518.2 | - | 9 | 845 | 40033360 | ||
BC143655.1 | - | 9 | 860 |
Set Membership: |
G-protein coupled receptor |