Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTBN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020971.2 | NP_066022.2 | 19 | 4118 | ||
NM_025213.2 | NP_079489.2 | 2 | 171 | |||
XM_011527173.2 | XP_011525475.1 | 19 | 4184 | |||
XM_017027049.1 | XP_016882538.1 | 19 | 4371 | |||
XM_017027050.1 | XP_016882539.1 | 17 | 3753 | |||
XM_017027051.1 | XP_016882540.1 | 19 | 4184 | |||
GenBank mRNA | AB046862.1 | - | 10 | 2861 | ||
AF082075.1 | - | 19 | 4179 | |||
AF311855.1 | - | 19 | 4118 | |||
AF311856.1 | - | 2 | 171 | |||
AF324063.1 | - | 19 | 4102 | |||
AF324064.1 | - | 19 | 4102 | |||
AK308547.1 | - | 2 | 159 | |||
AK309418.1 | - | 2 | 133 | |||
AY004226.1 | - | 19 | 4087 | |||
AY004227.1 | - | 18 | 4001 | |||
DA514440.1 | - | 2 | 133 |