Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172656.1 | NP_001166127.1 | 7 | 1098 | ||
NM_001172659.1 | NP_001166130.1 | 8 | 1221 | |||
NM_020972.2 | NP_066023.2 | 8 | 1188 | |||
XM_006713900.3 | XP_006713963.1 | 8 | 1242 | |||
XM_006713902.3 | XP_006713965.1 | 8 | 1243 | |||
XM_011513540.2 | XP_011511842.1 | 6 | 656 | |||
XM_011513541.2 | XP_011511843.1 | 8 | 1242 | |||
XM_017008502.1 | XP_016863991.1 | 8 | 2681 | |||
XM_017008503.1 | XP_016863992.1 | 8 | 1243 | |||
XM_017008504.1 | XP_016863993.1 | 8 | 1242 | |||
XM_017008505.1 | XP_016863994.1 | 7 | 2786 | |||
XM_017008506.1 | XP_016863995.1 | 8 | 1243 | |||
XM_017008507.1 | XP_016863996.1 | 8 | 1242 | |||
XM_017008508.1 | XP_016863997.1 | 8 | 1242 | |||
XM_017008509.1 | XP_016863998.1 | 8 | 1242 | |||
XM_017008510.1 | XP_016863999.1 | 8 | 1242 | |||
GenBank mRNA | AB046863.1 | - | 8 | 1187 | ||
AK126692.1 | - | 6 | 2053 | |||
AK293775.1 | - | 5 | 581 | |||
AK294710.1 | - | 8 | 925 | |||
AK294874.1 | - | 8 | 1149 | |||
AK316484.1 | - | 8 | 1221 | |||
BC137309.1 | - | 9 | 931 | 9021306 | ||
BC137310.1 | - | 9 | 931 | 9021307 |