Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031417.3 | NP_113605.2 | 16 | 2241 | ||
GenBank mRNA | AB049127.1 | - | 16 | 2207 | ||
AB058763.1 | - | 16 | 1915 | |||
BC009049.2 | - | 2 | 71 |