Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172231.3 | NP_757386.2 | 5 | 660 | ||
XM_005260002.2 | XP_005260059.1 | 4 | 675 | |||
XM_011528155.2 | XP_011526457.1 | 4 | 553 | |||
GenBank mRNA | AF521128.1 | - | 5 | 660 | ||
AK096152.1 | - | 4 | 749 | |||
AK292085.1 | - | 5 | 657 | |||
AK294618.1 | - | 5 | 722 | |||
AY072916.1 | - | 5 | 836 | |||
AY072917.1 | - | 5 | 640 | |||
BC063784.1 | - | 5 | 638 | |||
BC142988.1 | - | 5 | 657 | 8860435 |