Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB40C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172663.1 | NP_001166134.1 | 2, 3 | 364 | ||
NM_001172664.1 | NP_001166135.1 | 2, 3 | 256 | |||
NM_001172665.1 | NP_001166136.1 | 2, 3 | 216 | |||
NM_001172666.1 | NP_001166137.1 | 2, 3 | 216 | |||
NM_021168.4 | NP_066991.3 | 1, 2 | 261 | |||
GenBank mRNA | AY823398.1 | - | 1, 2 | 186 | ||
BC028696.1 | - | 1, 2 | 263 | 4826800 | ||
BG696681.1 | - | 2, 3 | 365 | |||
BG715678.1 | - | 2, 3 | 259 | |||
BM545442.1 | - | 1, 2 | 335 | |||
BQ685517.1 | - | 2, 3 | 195 | |||
CX783728.1 | - | 2, 3 | 349 |