Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021174.5 | NP_066997.3 | 19 | 2916 | ||
NR_033902.1 | 19 | 2734 | ||||
XM_011544603.1 | XP_011542905.1 | 19 | 2731 | |||
XM_011544604.1 | XP_011542906.1 | 18 | 2614 | |||
XM_017013717.1 | XP_016869206.1 | 19 | 2913 | |||
GenBank mRNA | AB075847.1 | - | 16 | 2225 | ||
AF293335.1 | - | 8 | 1171 | |||
AK022661.1 | - | 6 | 818 | |||
AK095712.1 | - | 19 | 2590 | |||
AK096547.1 | - | 10 | 1565 | |||
AK314535.1 | - | 19 | 2609 | |||
AL137523.1 | - | 5 | 544 | |||
AL834162.1 | - | 19 | 2903 | |||
AL834351.1 | - | 19 | 2904 | |||
AL834352.1 | - | 19 | 2904 | |||
BC018269.1 | - | 7 | 938 | |||
BC065495.1 | - | 19 | 2592 | 5496068 | ||
BX640952.1 | - | 19 | 2603 |