Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081686.1 | NP_001075155.1 | 1 | 161 | ||
NM_021176.2 | NP_066999.1 | 1 | 161 | |||
XM_011511564.2 | XP_011509866.1 | 1 | 178 | |||
GenBank mRNA | BC104778.1 | - | 1 | 161 | 8143781 | |
BC113376.1 | - | 1 | 161 | 8322428 | ||
BQ268961.1 | - | 1 | 96 | |||
CR627438.1 | - | 1 | 112 |