Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021178.4 | NP_067001.3 | 6 | 1042 | ||
NM_182849.2 | NP_878269.1 | 5 | 842 | |||
NM_182852.3 | NP_878272.1 | 7 | 1223 | |||
GenBank mRNA | AF216381.1 | - | 2 | 553 | ||
AK026233.1 | - | 6 | 1009 | |||
AK225253.1 | - | 6 | 1014 | |||
AK225451.1 | - | 6 | 1009 | |||
AK226037.1 | - | 6 | 1014 | |||
AL161994.1 | - | 6 | 1022 | |||
BC000369.2 | - | 5 | 984 | 2819346 | ||
BC001218.1 | - | 5 | 987 | 3528496 | ||
BC004435.1 | - | 5 | 984 | 2819346 | ||
BX161423.1 | - | 6 | 1202 | |||
BX161424.1 | - | 6 | 1170 | |||
BX247978.1 | - | 6 | 1150 |