Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021178.4 | NP_067001.3 | 6 | 992 | ||
NM_182849.2 | NP_878269.1 | 5 | 792 | |||
NM_182852.3 | NP_878272.1 | 7 | 1173 | |||
GenBank mRNA | AF216381.1 | - | 2 | 503 | ||
AK026233.1 | - | 6 | 959 | |||
AK225253.1 | - | 6 | 964 | |||
AK225451.1 | - | 6 | 959 | |||
AK226037.1 | - | 6 | 964 | |||
AL161994.1 | - | 6 | 972 | |||
BC000369.2 | - | 5 | 934 | 2819346 | ||
BC001218.1 | - | 5 | 937 | 3528496 | ||
BC004435.1 | - | 5 | 934 | 2819346 | ||
BX161423.1 | - | 6 | 1152 | |||
BX161424.1 | - | 6 | 1120 | |||
BX247978.1 | - | 6 | 1100 |