Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282589.1 | NP_001269518.1 | 3, 4 | 721 | ||
NM_001282590.1 | NP_001269519.1 | 4, 5 | 793 | |||
NM_001282591.1 | NP_001269520.1 | 3, 4 | 673 | |||
NM_001282594.1 | NP_001269523.1 | 4, 5 | 832 | |||
NM_001282595.1 | NP_001269524.1 | 4, 5 | 575 | |||
NM_021181.4 | NP_067004.3 | 5, 6 | 1114 | |||
XM_011509828.1 | XP_011508130.1 | 5, 6 | 963 | |||
GenBank mRNA | AB027233.1 | - | 5, 6 | 884 | ||
AF291815.1 | - | 5, 6 | 880 | |||
AF390894.1 | - | 5, 6 | 889 | |||
AJ276429.2 | - | 5, 6 | 902 | |||
AK292097.1 | - | 4, 5 | 577 | |||
AK292148.1 | - | 5, 6 | 898 | |||
AK298499.1 | - | 3, 4 | 528 | |||
AK301432.1 | - | 4, 5 | 618 | |||
AK301438.1 | - | 3, 4 | 461 | |||
AL834424.1 | - | 4, 5 | 582 | |||
AY358512.1 | - | 5, 6 | 906 | |||
DA978525.1 | - | 556 |