Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282588.1 | NP_001269517.1 | 3 | 654 | ||
NM_001282589.1 | NP_001269518.1 | 4 | 758 | |||
NM_001282590.1 | NP_001269519.1 | 5 | 830 | |||
NM_001282591.1 | NP_001269520.1 | 4 | 710 | |||
NM_001282592.1 | NP_001269521.1 | 5 | 1047 | |||
NM_001282593.1 | NP_001269522.1 | 3 | 606 | |||
NM_001282594.1 | NP_001269523.1 | 5 | 869 | |||
NM_001282595.1 | NP_001269524.1 | 5 | 612 | |||
NM_001282596.1 | NP_001269525.1 | 4 | 726 | |||
NM_021181.4 | NP_067004.3 | 6 | 1151 | |||
XM_011509828.1 | XP_011508130.1 | 6 | 1000 | |||
XM_011509829.1 | XP_011508131.1 | 5 | 896 | |||
GenBank mRNA | AB027233.1 | - | 6 | 921 | ||
AF291815.1 | - | 6 | 917 | |||
AF390894.1 | - | 6 | 926 | |||
AJ276429.2 | - | 6 | 939 | |||
AK290706.1 | - | 5 | 831 | |||
AK292097.1 | - | 5 | 614 | |||
AK292148.1 | - | 6 | 935 | |||
AK298499.1 | - | 4 | 565 | |||
AK298548.1 | - | 3 | 462 | |||
AK301137.1 | - | 3 | 397 | |||
AK301432.1 | - | 5 | 655 | |||
AK301438.1 | - | 4 | 498 | |||
AL713801.1 | - | 1 | 1776 | |||
AL834424.1 | - | 5 | 619 | |||
AY358512.1 | - | 6 | 943 | |||
BC027867.1 | - | 5 | 813 | 5203638 | ||
DA644251.1 | - | 4 | 447 |