Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021184.3 | NP_067007.3 | 1 | 1347 | ||
GenBank mRNA | AJ245433.1 | - | 1 | 1308 | ||
AK315349.1 | - | 1 | 1036 | |||
BC012950.1 | - | 1 | 1347 | 3875402 |