Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC31A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001859.3 | NP_001850.1 | 5 | 646 | ||
GenBank mRNA | AK129613.1 | - | 5 | 588 | ||
AK222866.1 | - | 5 | 588 | |||
AK292511.1 | - | 5 | 600 | |||
AL831843.1 | - | 5 | 586 | |||
BC004315.1 | - | 2 | 135 | |||
BC013611.1 | - | 5 | 575 | 3881191 | ||
U83460.1 | - | 5 | 612 |