Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC31A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001859.3 | NP_001850.1 | 2, 3 | 323 | ||
GenBank mRNA | AK129613.1 | - | 2, 3 | 265 | ||
AK222866.1 | - | 2, 3 | 265 | |||
AK292511.1 | - | 2, 3 | 277 | |||
AL831843.1 | - | 2, 3 | 263 | |||
BC013611.1 | - | 2, 3 | 252 | 3881191 | ||
BC061924.1 | - | 2, 3 | 251 | 4802875 | ||
U83460.1 | - | 2, 3 | 289 |