Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282466.1 | NP_001269395.1 | 7 | 1015 | ||
NM_001282467.1 | NP_001269396.1 | 7 | 1012 | |||
NM_021197.3 | NP_067020.2 | 7 | 1019 | |||
XM_005256079.3 | XP_005256136.1 | 6 | 930 | |||
GenBank mRNA | AF169631.1 | - | 7 | 907 | ||
AF302109.1 | - | 7 | 932 | |||
AK075061.1 | - | 7 | 875 | |||
AL713785.1 | - | 4 | 3677 | |||
BC029159.1 | - | 7 | 825 | 5186156 | ||
BQ670532.1 | - | 7 | 635 | |||
BQ671964.1 | - | 7 | 636 |