Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206878.1 | NP_001193807.1 | 4 | 415 | ||
NM_021198.2 | NP_067021.1 | 4 | 679 | |||
NM_182642.2 | NP_872580.1 | 4 | 676 | |||
XM_011511569.2 | XP_011509871.1 | 4 | 826 | |||
XM_011511570.2 | XP_011509872.1 | 4 | 822 | |||
XM_017004615.1 | XP_016860104.1 | 4 | 837 | |||
XM_017004616.1 | XP_016860105.1 | 4 | 832 | |||
XM_017004617.1 | XP_016860106.1 | 4 | 1261 | |||
XM_017004618.1 | XP_016860107.1 | 4 | 443 | |||
GenBank mRNA | AF217969.1 | - | 2 | 83 | ||
AF229162.1 | - | 4 | 679 | |||
AF258582.1 | - | 2 | 139 | |||
AY279529.1 | - | 4 | 340 | |||
AY279530.1 | - | 3 | 199 | |||
BC012977.2 | - | 4 | 388 | 3687816 | ||
BX424971.2 | - | 4 | 303 | |||
BX446444.2 | - | 4 | 413 |