Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206878.1 | NP_001193807.1 | 7 | 747 | ||
NM_021198.2 | NP_067021.1 | 7 | 1011 | |||
NM_182642.2 | NP_872580.1 | 7 | 1008 | |||
XM_011511569.2 | XP_011509871.1 | 7 | 1158 | |||
XM_011511570.2 | XP_011509872.1 | 7 | 1154 | |||
GenBank mRNA | AF217969.1 | - | 5 | 415 | ||
AF229162.1 | - | 7 | 1011 | |||
AF258582.1 | - | 5 | 471 | |||
AK075090.1 | - | 2 | 140 | |||
AY279529.1 | - | 7 | 672 | |||
AY279530.1 | - | 6 | 531 | |||
BC012977.2 | - | 7 | 720 | 3687816 | ||
BX424971.2 | - | 7 | 635 | |||
BX446444.2 | - | 7 | 745 |