Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100812.1 | NP_001094282.1 | 4 | 1083 | ||
NM_022059.3 | NP_071342.2 | 4 | 1083 | |||
GenBank mRNA | AF275260.1 | - | 4 | 594 | ||
AF301016.1 | - | 4 | 974 | |||
AF337812.1 | - | 4 | 508 | |||
AK027389.1 | - | 4 | 1001 | |||
AK292114.1 | - | 4 | 925 | |||
AK315017.1 | - | 4 | 1083 | |||
AY358909.1 | - | 4 | 928 | |||
BC017588.1 | - | 4 | 1077 | 4837010 | ||
BC044930.1 | - | 4 | 778 | 5762458 | ||
JC209418.1 | - | 4 | 1083 | |||
JC209420.1 | - | 4 | 1083 |