Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130033.1 | NP_001123505.1 | 3 | 660 | ||
NM_001130034.1 | NP_001123506.1 | 3 | 401 | |||
NM_001130035.1 | NP_001123507.1 | 3 | 401 | |||
NM_001130036.1 | NP_001123508.1 | 2 | 533 | |||
NM_021200.2 | NP_067023.1 | 3 | 660 | |||
XM_011545192.2 | XP_011543494.1 | 3 | 1234 | |||
XM_011545193.2 | XP_011543495.1 | 3 | 1234 | |||
GenBank mRNA | AB209716.1 | - | 3 | 313 | ||
AF081583.1 | - | 2 | 177 | |||
AF093249.1 | - | 3 | 285 | |||
AF100612.1 | - | 3 | 285 | |||
AF101054.1 | - | 3 | 656 | |||
AK289620.1 | - | 3 | 313 | |||
AK289909.1 | - | 3 | 279 | |||
AK290109.1 | - | 2 | 533 | |||
AK296033.1 | - | 3 | 295 | |||
AK301299.1 | - | 3 | 296 | |||
AK314748.1 | - | 3 | 257 | |||
BC008075.2 | - | 3 | 339 | 3050369 | ||
DC305763.1 | - | 3 | 401 | |||
U89715.1 | - | 3 | 656 |