Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENOPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292017.1 | NP_001278946.1 | 5 | 894 | ||
NM_021204.4 | NP_067027.1 | 5 | 923 | |||
NR_120457.1 | 4 | 691 | ||||
GenBank mRNA | AF113125.1 | - | 5 | 794 | ||
AF177286.1 | - | 3 | 360 | |||
AK022656.1 | - | 5 | 789 | |||
AL110167.1 | - | 1 | 24 | |||
AL518796.3 | - | 5 | 726 | |||
BC001317.1 | - | 4 | 509 | |||
BC065815.1 | - | 5 | 761 | 6526329 | ||
CR457141.1 | - | 5 | 573 |