Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199138.1 | NP_001186067.1 | 8 | 2890 | ||
NM_001199139.1 | NP_001186068.1 | 8 | 2773 | |||
NM_001302504.1 | NP_001289433.1 | 7 | 909 | |||
NM_021209.4 | NP_067032.3 | 8 | 2904 | |||
GenBank mRNA | AF376061.1 | - | 9 | 3118 | ||
AK095467.1 | - | 8 | 2890 | |||
AK292673.1 | - | 8 | 2875 | |||
AK314762.1 | - | 8 | 2875 | |||
AL389934.1 | - | 5 | 875 | |||
AY027787.1 | - | 8 | 2905 | |||
AY027788.1 | - | 7 | 910 | |||
AY032589.1 | - | 7 | 2664 | |||
AY035391.1 | - | 8 | 2773 | |||
AY358152.1 | - | 8 | 2852 | |||
BC031555.1 | - | 8 | 2860 | 5179909 |