Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREBZF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039618.2 | NP_001034707.1 | 1 | 1054 | ||
NR_028024.1 | 1 | 1054 | ||||
NR_028025.1 | 1 | 1054 | ||||
NR_028026.1 | 1 | 1054 | ||||
NR_028027.1 | 1 | 1054 | ||||
XM_006718642.3 | XP_006718705.1 | 1 | 1064 | |||
XM_011545195.2 | XP_011543497.1 | 1 | 1064 | |||
XM_017018086.1 | XP_016873575.1 | 1 | 1064 | |||
XM_017018087.1 | XP_016873576.1 | 1 | 1064 | |||
XM_017018088.1 | XP_016873577.1 | 1 | 1064 | |||
XM_017018089.1 | XP_016873578.1 | 1 | 1064 | |||
XM_017018090.1 | XP_016873579.1 | 1 | 1064 | |||
XM_017018091.1 | XP_016873580.1 | 1 | 1064 | |||
XM_017018092.1 | XP_016873581.1 | 1 | 1064 | |||
GenBank mRNA | AF039942.1 | - | 1 | 1002 | ||
AK313471.1 | - | 1 | 743 | |||
BC060807.1 | - | 1 | 1045 | 30345789 | ||
BC093796.1 | - | 1 | 633 | |||
BC093798.1 | - | 1 | 633 | |||
BQ720558.1 | - | 1 | 105 | |||
CA843387.1 | - | 1 | 114 | |||
DQ128105.1 | - | 1 | 572 |