Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021216.4 | NP_067039.1 | 3 | 839 | ||
XM_011527193.2 | XP_011525495.1 | 4 | 991 | |||
XM_017027078.1 | XP_016882567.1 | 4 | 901 | |||
XM_017027079.1 | XP_016882568.1 | 4 | 2771 | |||
XM_017027080.1 | XP_016882569.1 | 3 | 774 | |||
GenBank mRNA | AF269249.1 | - | 2 | 685 | ||
AK057019.1 | - | 1 | 180 | |||
BC014280.2 | - | 3 | 811 | 3840512 |