Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX4I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318786.1 | NP_001305715.1 | 5 | 605 | ||
NM_001318794.1 | NP_001305723.1 | 5 | 866 | |||
NM_001318797.1 | NP_001305726.1 | 5 | 449 | |||
NM_001861.4 | NP_001852.1 | 5 | 584 | |||
GenBank mRNA | AK027136.1 | - | 1 | 3707 | ||
AK311847.1 | - | 5 | 456 | |||
BC008704.1 | - | 5 | 442 | 2990066 | ||
BC021236.2 | - | 5 | 435 | 4309698 | ||
BC047869.1 | - | 3 | 2030 | |||
BC062437.1 | - | 5 | 463 | 6451700 | ||
BE615615.1 | - | 4 | 403 | |||
BE728651.1 | - | 5 | 446 | |||
BT019825.1 | - | 4 | 397 | |||
BU599266.1 | - | 443 | ||||
M21575.1 | - | 5 | 431 | |||
M34600.1 | - | 4 | 397 | |||
U90915.1 | - | 5 | 467 | |||
X54802.1 | - | 5 | 459 |