Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6G5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021221.2 | NP_067044.2 | 2 | 160 | ||
GenBank mRNA | AJ245417.1 | - | 1 | 879 | ||
AJ315545.1 | - | 2 | 160 | |||
AK289364.1 | - | 2 | 944 | |||
CF264683.1 | - | 3 | 193 | |||
HE864450.1 | - | 2 | 180 | |||
HE864451.1 | - | 1 | 327 |