Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021223.2 | NP_067046.1 | 2 | 148 | ||
XM_005249817.4 | XP_005249874.1 | 3 | 291 | |||
XM_011515463.2 | XP_011513765.1 | 3 | 292 | |||
XM_011515464.2 | XP_011513766.1 | 3 | 222 | |||
XM_011515465.2 | XP_011513767.1 | 3 | 222 | |||
XM_017012478.1 | XP_016867967.1 | 3 | 221 | |||
XM_017012479.1 | XP_016867968.1 | 3 | 229 | |||
GenBank mRNA | AK311869.1 | - | 3 | 163 | ||
BC027915.1 | - | 2 | 144 | 5223298 | ||
M94547.1 | - | 2 | 148 | |||
W17098.1 | - | 3 | 149 |