Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302807.1 | NP_001289736.1 | 4 | 570 | ||
NM_021225.4 | NP_067048.4 | 3 | 514 | |||
NR_126503.1 | 2 | 448 | ||||
GenBank mRNA | CD722615.1 | - | 3 | 514 | ||
CD723077.1 | - | 4 | 497 | |||
CD723628.1 | - | 1 | 13 | |||
CK429697.1 | - | 2 | 365 | |||
S83198.1 | - | 3 | 420 |