Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGAP22 SIRNA Assays ›
Gene Symbol:
ARHGAP22
Gene Name:
Rho GTPase activating protein 22
Gene Aliases:
RhoGAP2, RhoGap22
Chromosome Location:
Chr.10: 48429831 - 48656265 on Build GRCh38
UniGene ID:
Hs.655672
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s33881
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGAP22
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256024.1
NP_001242953.1
10
2312
NM_001256025.2
NP_001242954.1
10
2178
NM_001256026.1
NP_001242955.1
8
1818
NM_021226.3
NP_067049.2
10
2264
NR_045675.1
11
2763
XM_005270014.3
XP_005270071.1
8
1722
XM_011540002.1
XP_011538304.1
11
2863
XM_011540003.1
XP_011538305.1
11
2165
XM_011540004.1
XP_011538306.1
11
2815
XM_011540005.1
XP_011538307.1
8
1866
XM_011540006.1
XP_011538308.1
10
2686
XM_011540007.2
XP_011538309.1
8
6070
XM_011540011.2
XP_011538313.1
7
1545
XM_011540012.2
XP_011538314.1
8
2223
XM_011540015.2
XP_011538317.1
10
1983
XM_017016470.1
XP_016871959.1
9
2132
XM_017016471.1
XP_016871960.1
8
2137
XM_017016472.1
XP_016871961.1
8
1710
XM_017016474.1
XP_016871963.1
10
2728
XM_017016475.1
XP_016871964.1
9
2599
XM_017016477.1
XP_016871966.1
10
2209
GenBank mRNA
AK055037.1
-
9
1857
AK127586.1
-
8
1818
AK293579.1
-
10
2064
AY324801.1
-
10
2226
BC047096.1
-
10
2264
BC126444.1
-
10
2133
8992160
BC136319.1
-
10
2133
9020306
EF560749.1
-
10
2236
U90908.1
-
2
1317
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
signal transduction
cell differentiation
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activator activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline