Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM60A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135811.1 | NP_001129283.1 | 6 | 731 | ||
NM_001135812.1 | NP_001129284.1 | 5 | 627 | |||
NM_021238.2 | NP_067061.1 | 6 | 727 | |||
XM_011520803.2 | XP_011519105.1 | 5 | 729 | |||
XM_017019758.1 | XP_016875247.1 | 6 | 795 | |||
XM_017019759.1 | XP_016875248.1 | 7 | 841 | |||
XM_017019760.1 | XP_016875249.1 | 7 | 846 | |||
XM_017019761.1 | XP_016875250.1 | 6 | 776 | |||
XM_017019762.1 | XP_016875251.1 | 6 | 693 | |||
GenBank mRNA | AF087885.1 | - | 6 | 680 | ||
AF212220.1 | - | 5 | 586 | |||
AK026932.1 | - | 5 | 538 | |||
AK294457.1 | - | 5 | 640 | |||
AL157432.1 | - | 6 | 679 | |||
AY598323.1 | - | 4 | 405 | |||
BC000024.2 | - | 6 | 727 | 3504430 | ||
BC004462.1 | - | 2 | 143 | |||
BC071966.1 | - | 5 | 507 | 5924274 | ||
BX648630.1 | - | 4 | 722 | |||
DR763430.1 | - | 2 | 138 |