Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021259.2 | NP_067082.2 | 13 | 2266 | ||
GenBank mRNA | AB045292.1 | - | 13 | 2223 | ||
AK024725.1 | - | 10 | 1565 | |||
AK091823.1 | - | 13 | 2044 | |||
BC004276.1 | - | 3 | 291 | |||
BC021557.1 | - | 13 | 2157 | 4899167 |