Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290072.1 | NP_001277001.1 | 5 | 1325 | ||
NM_001290073.1 | NP_001277002.1 | 4 | 972 | |||
NM_021572.5 | NP_067547.1 | 4 | 1263 | |||
XM_005249260.3 | XP_005249317.1 | 4 | 1162 | |||
XM_011514786.2 | XP_011513088.1 | 5 | 1180 | |||
GenBank mRNA | AK292842.1 | - | 4 | 1224 | ||
AK295192.1 | - | 4 | 933 | |||
AK312790.1 | - | 4 | 1105 | |||
AY358514.1 | - | 5 | 1268 | |||
BC027615.2 | - | 4 | 1163 | 4839342 | ||
BX647968.1 | - | 4 | 1234 |