Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286556.1 | NP_001273485.1 | 8 | 1667 | ||
NM_001320744.1 | NP_001307673.1 | 6 | 1411 | |||
NM_001320745.1 | NP_001307674.1 | 8 | 1601 | |||
NM_058187.4 | NP_478067.2 | 8 | 1676 | |||
NR_104472.1 | 7 | 1470 | ||||
NR_135467.1 | 8 | 1775 | ||||
XM_005261019.4 | XP_005261076.1 | 6 | 1626 | |||
XM_006724038.3 | XP_006724101.1 | 8 | 1794 | |||
XM_011529669.2 | XP_011527971.1 | 8 | 1928 | |||
XM_017028418.1 | XP_016883907.1 | 8 | 1794 | |||
XM_017028419.1 | XP_016883908.1 | 7 | 1716 | |||
XM_017028420.1 | XP_016883909.1 | 9 | 1717 | |||
XM_017028421.1 | XP_016883910.1 | 8 | 1592 | |||
XM_017028422.1 | XP_016883911.1 | 6 | 1467 | |||
XM_017028423.1 | XP_016883912.1 | 5 | 1395 | |||
GenBank mRNA | AF358258.1 | - | 8 | 1259 | ||
AK125013.1 | - | 7 | 2226 | |||
AY040087.1 | - | 7 | 1062 | |||
AY358787.1 | - | 8 | 1669 | |||
BC038710.1 | - | 8 | 1402 | 4813873 |