Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021229.3 | NP_067052.2 | 10 | 2228 | ||
GenBank mRNA | AF119916.3 | - | 11 | 1765 | ||
AF278532.1 | - | 10 | 2230 | |||
AF297711.1 | - | 10 | 2080 | |||
AK024691.1 | - | 9 | 1304 | |||
AK026833.1 | - | 5 | 574 | |||
AK226070.1 | - | 10 | 2085 | |||
AK290591.1 | - | 10 | 1765 | |||
AK315796.1 | - | 10 | 2087 | |||
AL137540.1 | - | 8 | 1096 | |||
AL833767.1 | - | 9 | 1632 | |||
AY330211.1 | - | 9 | 1710 | |||
BC013591.1 | - | 9 | 1303 | |||
BC136798.1 | - | 10 | 2203 | 9020788 | ||
BC136799.1 | - | 10 | 2203 | 9020789 |