Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031897.2 | NP_114103.2 | 2 | 1602 | ||
NM_145814.1 | NP_665813.1 | 4 | 1815 | |||
NM_145815.1 | NP_665814.1 | 3 | 1677 | |||
NR_102308.1 | 3 | 805 | ||||
GenBank mRNA | BC021552.1 | - | 4 | 1815 | 3357889 |