Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004075.4 | NP_004066.1 | 6 | 1611 | ||
XM_017018832.1 | XP_016874321.1 | 7 | 1746 | |||
GenBank mRNA | AK098615.1 | - | 1 | 14 | ||
AK125915.1 | - | 8 | 1274 | |||
AK290552.1 | - | 6 | 911 | |||
BC017466.1 | - | 6 | 1034 | |||
BC030519.1 | - | 6 | 1284 | 5169081 | ||
D83702.1 | - | 6 | 1301 | |||
D84657.1 | - | 6 | 715 |