Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX7A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001864.3 | NP_001855.1 | 3 | 582 | ||
GenBank mRNA | AK312091.1 | - | 3 | 421 | ||
BC002757.2 | - | 3 | 582 | 3631740 | ||
BT006924.1 | - | 3 | 120 | |||
CR542120.1 | - | 3 | 120 | |||
HE647857.1 | - | 3 | 117 | |||
M83186.1 | - | 3 | 161 |