Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177428.1 | NP_001170899.1 | 4 | 914 | ||
NM_001177431.1 | NP_001170902.1 | 6 | 922 | |||
NM_001177433.1 | NP_001170904.1 | 4 | 914 | |||
NM_021625.4 | NP_067638.3 | 6 | 1113 | |||
NM_147204.2 | NP_671737.1 | 5 | 1055 | |||
XM_005253918.1 | XP_005253975.1 | 6 | 1473 | |||
XM_011538630.2 | XP_011536932.2 | 6 | 2216 | |||
XM_011538631.2 | XP_011536933.2 | 5 | 2076 | |||
XM_011538632.2 | XP_011536934.2 | 6 | 2217 | |||
XM_011538633.2 | XP_011536935.2 | 5 | 2076 | |||
XM_011538634.2 | XP_011536936.2 | 6 | 2218 | |||
XM_011538635.2 | XP_011536937.1 | 6 | 2220 | |||
XM_011538636.2 | XP_011536938.1 | 6 | 2221 | |||
XM_017019774.1 | XP_016875263.1 | 6 | 1846 | |||
GenBank mRNA | AB032427.1 | - | 5 | 1119 | ||
AB073669.1 | - | 5 | 1024 | |||
AB100308.1 | - | 5 | 1024 | |||
AF258465.1 | - | 5 | 1024 | |||
AF263523.1 | - | 5 | 1024 | |||
AF279673.1 | - | 5 | 820 | |||
AJ296305.1 | - | 6 | 1113 | |||
AL834192.1 | - | 6 | 1086 | |||
BC117426.1 | - | 6 | 1107 | 40125977 | ||
BC143307.1 | - | 6 | 1107 | |||
BC143315.1 | - | 6 | 1107 | 9051820 | ||
DQ059644.1 | - | 4 | 883 | |||
DQ059645.1 | - | 6 | 922 | |||
DQ059646.1 | - | 4 | 883 |