Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177431.1 | NP_001170902.1 | 5 | 658 | ||
NM_021625.4 | NP_067638.3 | 5 | 849 | |||
NM_147204.2 | NP_671737.1 | 4 | 791 | |||
XM_005253918.1 | XP_005253975.1 | 5 | 1209 | |||
XM_011538630.2 | XP_011536932.2 | 5 | 1952 | |||
XM_011538632.2 | XP_011536934.2 | 5 | 1953 | |||
XM_011538634.2 | XP_011536936.2 | 5 | 1954 | |||
XM_011538635.2 | XP_011536937.1 | 5 | 1956 | |||
XM_011538636.2 | XP_011536938.1 | 5 | 1957 | |||
XM_017019774.1 | XP_016875263.1 | 5 | 1582 | |||
GenBank mRNA | AB032427.1 | - | 4 | 855 | ||
AB073669.1 | - | 4 | 760 | |||
AB100308.1 | - | 4 | 760 | |||
AF258465.1 | - | 4 | 760 | |||
AF263523.1 | - | 4 | 760 | |||
AF279673.1 | - | 4 | 556 | |||
AJ296305.1 | - | 5 | 849 | |||
AL834192.1 | - | 5 | 821 | |||
BC117426.1 | - | 5 | 843 | 40125977 | ||
BC143307.1 | - | 5 | 843 | |||
BC143315.1 | - | 5 | 843 | 9051820 | ||
DQ059645.1 | - | 5 | 658 |