Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021627.2 | NP_067640.2 | 13 | 1592 | ||
XM_005247690.2 | XP_005247747.2 | 12 | 1475 | |||
XM_005247691.2 | XP_005247748.1 | 13 | 1504 | |||
XM_017006973.1 | XP_016862462.1 | 12 | 1372 | |||
GenBank mRNA | AF151697.2 | - | 13 | 1595 | ||
AK027599.1 | - | 13 | 1533 | |||
AK074357.1 | - | 8 | 963 | |||
AK296744.1 | - | 10 | 1137 | |||
AK296772.1 | - | 12 | 1473 | |||
AK298628.1 | - | 13 | 1419 | |||
AK304402.1 | - | 13 | 1661 | |||
AL834380.1 | - | 13 | 1416 | |||
BC040609.1 | - | 13 | 1478 | 5163937 |