Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOXE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165960.1 | NP_001159432.1 | 3 | 812 | ||
NM_021628.2 | NP_067641.2 | 3 | 648 | |||
XM_017024921.1 | XP_016880410.1 | 2 | 591 | |||
XM_017024922.1 | XP_016880411.1 | 2 | 591 | |||
XM_017024923.1 | XP_016880412.1 | 2 | 591 | |||
XM_017024924.1 | XP_016880413.1 | 2 | 591 | |||
XM_017024925.1 | XP_016880414.1 | 2 | 591 | |||
GenBank mRNA | AF182218.1 | - | 2 | 479 | ||
AJ269499.1 | - | 2 | 777 | |||
AK292732.1 | - | 2 | 551 | |||
AK296416.1 | - | 3 | 713 | |||
AK299742.1 | - | 3 | 755 | |||
AK313677.1 | - | 3 | 612 | |||
BC101938.1 | - | 2 | 500 | 40011855 | ||
BC101939.1 | - | 2 | 500 | 40011856 | ||
BC103508.1 | - | 2 | 500 | 40011857 | ||
BC104724.1 | - | 2 | 500 | 40011849 |