Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021640.3 | NP_067653.3 | 2 | 370 | ||
GenBank mRNA | AF289485.1 | - | 2 | 338 | ||
AK293449.1 | - | 2 | 326 | |||
BC013956.2 | - | 2 | 310 | |||
BC028904.1 | - | 1 | 274 | 4454943 | ||
BC051871.1 | - | 2 | 329 | 5170816 |