Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037126.1 | NP_001032203.1 | 8 | 1274 | ||
NM_021807.3 | NP_068579.3 | 8 | 1274 | |||
XM_005250523.4 | XP_005250580.1 | 8 | 1258 | |||
XM_017012495.1 | XP_016867984.1 | 8 | 1258 | |||
GenBank mRNA | AB051486.1 | - | 8 | 1216 | ||
AF132734.1 | - | 2 | 230 | |||
AF380839.1 | - | 8 | 1271 | |||
AK022751.1 | - | 1 | 42 | |||
AK027688.1 | - | 8 | 1268 | |||
AK295404.1 | - | 8 | 1282 | |||
AK298191.1 | - | 3 | 256 | |||
AL831989.2 | - | 8 | 1269 | |||
AL834475.1 | - | 5 | 601 | |||
BC020607.2 | - | 2 | 233 | |||
BC026174.1 | - | 8 | 1250 | |||
BC067263.1 | - | 8 | 1251 | 6183004 |