Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGIF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199513.1 | NP_001186442.1 | 3 | 349 | ||
NM_001199514.1 | NP_001186443.1 | 3 | 477 | |||
NM_001199515.1 | NP_001186444.1 | 3 | 379 | |||
NM_021809.6 | NP_068581.1 | 3 | 436 | |||
GenBank mRNA | AB042646.1 | - | 3 | 445 | ||
AK074802.1 | - | 3 | 416 | |||
AK313919.1 | - | 3 | 436 | |||
AL705108.1 | - | 3 | 349 | |||
BC006549.2 | - | 3 | 423 | 2964507 | ||
BC012816.2 | - | 3 | 423 | 2964507 | ||
BG723865.1 | - | 3 | 464 | |||
DA438831.1 | - | 3 | 436 |