Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021810.4 | NP_068582.2 | 2 | 294 | ||
NM_177980.2 | NP_817089.1 | 14 | 2397 | |||
XM_011528969.2 | XP_011527271.1 | 14 | 2403 | |||
XM_011528970.2 | XP_011527272.1 | 8 | 1419 | |||
XM_017027994.1 | XP_016883483.1 | 14 | 3066 | |||
XM_017027995.1 | XP_016883484.1 | 14 | 3067 | |||
XM_017027996.1 | XP_016883485.1 | 14 | 2968 | |||
XM_017027997.1 | XP_016883486.1 | 14 | 3067 | |||
XM_017027998.1 | XP_016883487.1 | 14 | 3064 | |||
XM_017027999.1 | XP_016883488.1 | 14 | 3064 | |||
XM_017028000.1 | XP_016883489.1 | 13 | 2236 | |||
XM_017028001.1 | XP_016883490.1 | 14 | 3064 | |||
XM_017028002.1 | XP_016883491.1 | 14 | 3065 | |||
GenBank mRNA | AF169690.1 | - | 6 | 861 | ||
AK055202.1 | - | 14 | 2386 | |||
AK074477.1 | - | 2 | 282 | |||
BC062570.1 | - | 2 | 294 | |||
BC136306.1 | - | 14 | 2201 | 9020293 | ||
BC143931.1 | - | 14 | 2201 |