Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242828.1 | NP_001229757.1 | 7 | 1013 | ||
NM_001301856.1 | NP_001288785.1 | 6 | 1165 | |||
NM_021814.4 | NP_068586.1 | 6 | 932 | |||
GenBank mRNA | AF231981.1 | - | 5 | 561 | ||
AF338241.1 | - | 6 | 905 | |||
AK074748.1 | - | 1 | 29 | |||
AK074889.1 | - | 5 | 566 | |||
AK125098.1 | - | 6 | 1009 | |||
AK299674.1 | - | 6 | 673 | |||
AK302948.1 | - | 7 | 784 | |||
AL136939.1 | - | 6 | 680 | |||
BC067123.1 | - | 6 | 681 | 6160926 |