Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303.3 | NP_001294.2 | 3 | 646 | ||
GenBank mRNA | AK295925.1 | - | 3 | 444 | ||
AK312718.1 | - | 3 | 541 | |||
BC000060.2 | - | 3 | 519 | 3508180 | ||
BC006394.2 | - | 3 | 519 | 4136163 | ||
BT006985.1 | - | 3 | 442 | |||
U09466.1 | - | 3 | 482 |