Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031712.2 | NP_001026882.2 | 12 | 1301 | ||
XM_017011168.1 | XP_016866657.1 | 11 | 1109 | |||
XM_017011169.1 | XP_016866658.1 | 11 | 1271 | |||
GenBank mRNA | AF182423.1 | - | 14 | 1431 | ||
AF532977.1 | - | 12 | 1266 | |||
BC056878.1 | - | 12 | 1301 | 4443778 |