Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC90B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286116.1 | NP_001273045.1 | 2 | 404 | ||
NM_001286117.1 | NP_001273046.1 | 2 | 951 | |||
NM_001286118.1 | NP_001273047.1 | 2 | 631 | |||
NM_001286119.1 | NP_001273048.1 | 3 | 716 | |||
NM_021825.4 | NP_068597.2 | 2 | 631 | |||
XM_005274154.2 | XP_005274211.1 | 2 | 890 | |||
XM_005274156.3 | XP_005274213.1 | 3 | 722 | |||
XM_005274159.3 | XP_005274216.1 | 3 | 297 | |||
XM_017018103.1 | XP_016873592.1 | 2 | 333 | |||
XM_017018104.1 | XP_016873593.1 | 3 | 722 | |||
GenBank mRNA | AF182412.1 | - | 2 | 201 | ||
AF182424.1 | - | 2 | 201 | |||
AF271782.1 | - | 2 | 558 | |||
AK055972.1 | - | 2 | 948 | |||
AK225726.1 | - | 2 | 212 | |||
AK304769.1 | - | 2 | 364 | |||
AK315787.1 | - | 2 | 592 | |||
AL136791.1 | - | 2 | 351 | |||
AV756714.1 | - | 2 | 271 | |||
BC014573.1 | - | 2 | 241 | 3680689 | ||
BC020783.1 | - | 2 | 202 | 4773466 | ||
BC067764.1 | - | 2 | 522 | |||
BC107755.1 | - | 2 | 372 | 6737596 | ||
CN309108.1 | - | 2 | 631 |